Order Entry
Puerto Rico
ContactUsLinkComponent
3,4,5-Trifluorobenzonitrile 97%
3,4,5-Trifluorobenzonitrile 97%
Catalog # 77417-720
Supplier:  APOLLO SCIENTIFIC
CAS Number:  
3,4,5-Trifluorobenzonitrile 97%
Catalog # 77417-720
Supplier:  APOLLO SCIENTIFIC
Supplier Number:  PC7283T-25G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    25 g
  • Cat. No.:
    77417-720

Specifications

About this item

 :
 :157.09 g/mol
Boiling Pt:72…73 °C/10 mmHg
Melting Pt:45…47 °C
Flash Pt:63 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
 :GHS07
CAS Number:134227-45-5
Formula:C₇H₂F₃N
MDL Number:MFCD00074948