VWR® Waugh Forceps
203 mm (8")
Catalog # 82027-428
Supplier: VWR International
Jump to:
Product Details & Documents
Waugh-type forcep with 1x2 teeth and guide pin.
- Stainless steel construction
- Waugh-type design
- Teeth help ensure a firm grip
Forceps are straight and feature cross-serrated tips.
Specifications
- Length:203 mm (8")