ContactUsLinkComponent
VWR® Hemostatic Forceps, Serrated Tip
VWR® Hemostatic Forceps, Serrated Tip
Catalog # 82027-416
Supplier:  VWR International
undefined
VWR® Hemostatic Forceps, Serrated Tip
Catalog # 82027-416
Supplier:  VWR International
Specifications
  • Length:
    140 mm (5½")
  • Cat. No.:
    82027-416
Specifications
About this item
Hemostatic forceps are used in many surgical procedures, usually to control bleeding or the flow of liquids in tubing.
  • Stainless steel fabrication
  • Serrated tips
These forceps have ring handles just like scissors, and ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure.