Order Entry
Puerto Rico
ContactUsLinkComponent
4-Boronobenzenesulfonamide pinacol ester 98%
4-Boronobenzenesulfonamide pinacol ester 98%
Catalog # 77405-804
Supplier:  APOLLO SCIENTIFIC
CAS Number:  
4-Boronobenzenesulfonamide pinacol ester 98%
Catalog # 77405-804
Supplier:  APOLLO SCIENTIFIC
Supplier Number:  OR16633-500MG
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    500 mg
  • Cat. No.:
    77405-804

Specifications

About this item

 :
 :283.1516 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:214360-51-7
Formula:C₁₂H₁₈BNO₄S
MDL Number:MFCD06657806