Order Entry
Puerto Rico
ContactUsLinkComponent
1,1,1,5,5,5-Hexafluoroacetylacetone 98%
Flammable
Corrosive
Toxic
1,1,1,5,5,5-Hexafluoroacetylacetone 98%
Catalog # 77415-632
Supplier:  APOLLO SCIENTIFIC
CAS Number:  
Flammable
Corrosive
Toxic
1,1,1,5,5,5-Hexafluoroacetylacetone 98%
Catalog # 77415-632
Supplier:  APOLLO SCIENTIFIC
Supplier Number:  PC4590-25G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Nalgene bottle
  • Size:
    25 g
  • Cat. No.:
    77415-632

Specifications

About this item

 :
Boiling Pt:70 °C
Density:1.486 g/cm³ (25 °C)
Flash Pt:32 °C
ADR:8+3,II
 :208.06 g/mol
EINECS:216-191-0
UN:2920
 :GHS02
 :GHS05
 :GHS06
Storage Temperature:Ambient
CAS Number:1522-22-1
Formula:C₅H₂F₆O₂
MDL Number:MFCD00000426