Order Entry
Puerto Rico
ContactUsLinkComponent
Chloropentafluorobenzene 99%
Toxic
Chloropentafluorobenzene 99%
Catalog # 77413-462
Supplier:  APOLLO SCIENTIFIC
CAS Number:  
Toxic
Chloropentafluorobenzene 99%
Catalog # 77413-462
Supplier:  APOLLO SCIENTIFIC
Supplier Number:  PC2010-5G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    5 g
  • Cat. No.:
    77413-462

Specifications

About this item

 :
 :202.51 g/mol
Boiling Pt:122…123 °C/750 mmHg
Density:1.568 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:206-450-6
UN:2810
ADR:6.1,III
 :GHS06
CAS Number:344-07-0
Formula:C₆ClF₅
MDL Number:MFCD00000534