Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
3-Morpholinobenzaldehyde 98%

3-Morpholinobenzaldehyde 98%

Irritant
Supplier:   APOLLO SCIENTIFIC
3-Morpholinobenzaldehyde 98%
Supplier:  APOLLO SCIENTIFIC
Certificates
3-Morpholinobenzaldehyde 98%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:446866-87-1
  • Formula:C₁₁H₁₃NO₂
  • MDL Number:MFCD07772811
  • Molecular Weight:191.2264 g/mol
  • Melting Pt:68…70 °C
  • Storage Temperature:Ambient
  • UN:2811
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $158.46
    $158.46
    Each(1/Itm/Bx)
       $158.46

    Supplier Number: OR9592-1G
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $507.26
    $507.26
    Each(1/Itm/Bx)
       $507.26

    Supplier Number: OR9592-5G

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.