Trityl chloride 95%
: Trt-Cl, Chlorotriphenylmethane, Triphenylmethyl chloride, (Chloromethanetriyl)tribenzene, Triphenylchloromethane
:76-83-5
:
:C₁₉H₁₅Cl
:MFCD00000813
:374
:278.78 g/mol
:111…113 °C
:Ambient
:3261
:8 III
:GHS05
:GHS09
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.