Order Entry
Puerto Rico
ContactUsLinkComponent
[(5-Methyl-2-thienyl)methyl]amine hydrochloride 97%
Toxic
[(5-Methyl-2-thienyl)methyl]amine hydrochloride 97%
Catalog # 77404-286
Supplier:  APOLLO SCIENTIFIC
CAS Number:  
Toxic
[(5-Methyl-2-thienyl)methyl]amine hydrochloride 97%
Catalog # 77404-286
Supplier:  APOLLO SCIENTIFIC
Supplier Number:  OR12377-100MG
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    100 mg
  • Cat. No.:
    77404-286

Specifications

About this item

 :
Melting Pt:191…193 °C
 :163.67 g/mol
 :GHS06
Storage Temperature:Ambient
CAS Number:171661-55-5
Formula:C₆H₁₀ClNS
MDL Number:MFCD09817548