Order Entry
Puerto Rico
ContactUsLinkComponent
Pyridine-[D₅] (>99.50% D)
Flammable
Irritant
Pyridine-[D₅] (>99.50% D)
Catalog # 77401-322
Supplier:  APOLLO SCIENTIFIC
CAS Number:  
Flammable
Irritant
Pyridine-[D₅] (>99.50% D)
Catalog # 77401-322
Supplier:  APOLLO SCIENTIFIC
Supplier Number:  DE85D-1ML
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass ampoule
  • Size:
    1 ml
  • Cat. No.:
    77401-322

Specifications

About this item

 :
 :84.06 g/mol
Boiling Pt:115 °C/570 mmHg
Melting Pt:41.99 °C
Density:1.05 g/cm³ (25 °C)
Flash Pt:17 °C
Storage Temperature:Ambient
EINECS:230-720-2
UN:1282
ADR:3,II
 :GHS02
 :GHS07
CAS Number:7291-22-7
Formula:C₅D₅N
MDL Number:MFCD00044639