Aquatic Block™ Serum-Free Tris (with Surfactant)
Catalog # 77395-196
Supplier: Arlington Scientific
CAS Number:
Specifications
- Description:Aquatic Block™ serum-free Tris (with surfactant), 500 ml
- Cat. No.:77395-196
Specifications
About this item
A proprietary blocking reagent supplied in tris for diagnostic systems involving membrane or other solid phase material.
- Imparts superior stability and flow characteristics
- Utilizes proteins that do not interact with mammalian antibodies
- Supplied in HDPE plastic
Preservative - 0.09% sodium azide, with surfactant.
Applications: Lateral flow, ELISA.