Order Entry
United States
ContactUsLinkComponent
Aquatic Block™ PBS, Azide-Free
Aquatic Block™ PBS, Azide-Free
Catalog # 77395-168
CAS Number:  
undefined
Aquatic Block™ PBS, Azide-Free
Catalog # 77395-168
Supplier Number:  PP71P-500
CAS Number:  

Specifications

  • Description:
    Aquatic Block™ PBS, azide-free, 500 ml
  • Environmentally Preferable:
  • Cat. No.:
    77395-168

Specifications

About this item

A non-mammalian blocking reagent supplied in PBS for diagnostic systems involving membrane or other solid phase material.

  • Optimized for superior performance in super sensitive assay systems
  • Utilizes proteins that do not interact with mammalian antibodies
  • Supplied in HDPE plastic

Azide-free.

Applications: ELISA/EIA/Western blot.