Blocking Buffer Sample Pack
Blocking buffer sample pack (6×100 ml)
Catalog # 77395-166
Supplier: Arlington Scientific
Supplier #: PP100-SAMPLEPK
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Contains: Aquatic Block PBS, Aquatic Block Tris, Aquatic Block Serum Free PBS, Aquatic Block Serum Free Tris,
Balance Blocker PBS, Balance Blocker Tris.
- Ready to use blocking buffer sample pack
- Supplied in HDPE plastic
Applications: ELISA, EIA, Western blot, Lateral flow, Nitrocellulose in lateral flow.
Specifications
Specifications
- Description:Blocking buffer sample pack (6×100 ml)



