12-channel reservoir plates are suitable for use as collection plates from filter systems, and are perfect for storing volumes of samples to be pipetted into other microplates for further applications. Standard SBS footprint ensures its compatibility with automation.
- Stackable
- Free from RNase, DNase, human genomic DNA, and endotoxins
- SBS footprint
- Compatible with plate readers and ideal for use with automation
- Suitable as a collection plate from filter systems
- Temperature range: −80 to +120 °C
Made from polypropylene, a material with very low biomolecular binding characteristics, a high temperature tolerance, and resistance to many standard laboratory chemicals, including DMSO. Reservoir plates feature a pyramid channel bottom, perfect for maximum sample retrieval. Both the open format plate (76638-054) and the 12-channel plate (76638-114) have pyramid bases. Rectangular channels (76638-114) ensure the best use of space and allow the use of multichannel pipettes. Plates are perfect for maximum sample retrieval, as even the smallest volume can be retrieved as it gathers in the base of the pyramid, and can be easily taken in by a pipette.