Nalgene® Polypropylene Locking Forceps, Thermo Scientific
30049-122
- Description:Plastic Forceps
- Cat. No.:30049-122
- Supplier no.:6320-0010
All-plastic forceps with double-action hinges for extra leverage and long life
Scissors-type handles with ratchet for locking tips shut. Interlocking teeth for tight grip. Provide a useful clamp for chromatography columns. Durable blue polypropylene is autoclavable for repeated use. Ideal for economical, disposable applications.