ContactUsLinkComponent
Rochester-Ochsner Hemostatic Forceps, Walter Stern
Rochester-Ochsner Hemostatic Forceps, Walter Stern
Catalog # 25607-380
Supplier:  Walter Stern
undefined
Rochester-Ochsner Hemostatic Forceps, Walter Stern
Catalog # 25607-380
Supplier:  Walter Stern
Supplier Number:  306-030
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Length:
    140 mm (5½")
  • Cat. No.:
    25607-380
  • Supplier no.:
    306-030
Specifications
About this item
Stainless steel
Rochester-Ochsner type. Straight blade. Serrated jaws with 1x2 teeth.