Orders LinkContactUsLinkComponent
Xcelite® Pliers, Cooper Tools
Xcelite® Pliers, Cooper Tools
Supplier:   Cooper Tools
Xcelite® Pliers, Cooper Tools
Supplier:  Cooper Tools
Certificates
Xcelite® Pliers, Cooper Tools
Xcelite® Pliers, Cooper Tools
2 Options Available Below
Product Details & Documents
Precision manufactured to handle most electronic assembly requirements, these pliers feature a steel rivet, ESD-safe cushion grips, and smooth, corrosion-proof joints.
Product Family Options
Product Information
  • Length
    Jaw type
    Jaw width
    Availability
    Price
  • 37693-472
    60° Thin-Tip Curved Nose Pliers
    60° Thin-Tip Curved Nose Pliers
    127 mm (5")
    Serrated
    31 mm (1⁷⁄₃₂")
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $205.27
    $205.27
    Log in for availability
    Pack of 6
    Pack of 6 - $1,231.76
    $1,231.76
    Each
       $205.27
    Pack of 6
       $1,231.76
    Supplier Number: CN255VN
  • 37693-474
    60° Thin-Tip Curved Nose Pliers
    60° Thin-Tip Curved Nose Pliers
    127 mm (5")
    Smooth
    31 mm (1⁷⁄₃₂")
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $1,283.64
    $1,283.64
    Log in for availability
    Pack of 6
    Pack of 6 - $7,701.73
    $7,701.73
    Each
       $1,283.64
    Pack of 6
       $7,701.73
    Supplier Number: CN255GN