About this item
These compact, flat shakers feature electronic adjustment of the speed and timer.
- 230 V model
The basic model has a timer with a range of 5 to 50 minutes. The control model features a timer with a range of 0 to 9 hours, 59 minutes, as well as a digital display to allow users to read the speed, timer function, and operating modes at a glance. Control model also features integrated end point positioning for automated robot-controlled sampling. All functions on the control model can be operated and documented with labworldsoft® software using the RS-232 interface.
The universal platform holds various types of vessels, and features variable clamping rolls that allow for universal adaptation. Universal platform surface dimensions: 32×32 cm (12¹/₂×12¹/₂"). The flask clamp platform is for shaking flasks, Erlenmeyer flasks, and bottles with a round cross-section. Accommodates fifty-six 25mL flasks, twenty-three 50 or 100 ml flasks, eleven 200 or 250 ml flasks, or nine 500 mL flasks. Flask clamps are available separately. Flask clamp platform surface dimensions: 33×33 cm (13×13"). The dish platform is for smooth movement of cell cultures, nutrient media, Petri dishes, culture bottles, and vessels with a low center of gravity. Dish platform surface dimensions: 32×32 cm (12¹/₂×12¹/₂"). The separating funnel platform is for shaking out, salting out, extracting, eluting, and enriching in pear-shaped separating funnels. Accommodates six 50mL funnels, five 100 ml funnels, three 250 ml funnels, and three 500 ml funnels. The three included clamping rolls are height-adjustable for adaptation to different separating funnel sizes. Separating funnels are secured with five included O-rings. Separating funnel platform surface dimensions: 33.4×42.5 cm (13¹/₈×16³/₄").
A wide range of attachment combinations makes it possible to use almost all vessel sizes and shapes.
Specifications
- Catalog No:
- 33998-366
- 33998-370
- Dimensions:
- 36W x 42D x 10H cm (143/16 x 1617/32 x 315/16")
- 36W x 42D x 10H cm (143/16 x 1617/32 x 315/16")
- Max. Load:
- 7.5 kg (16.5 lbs.)
- 7.5 kg (16.5 lbs.)
- Operating Temperature Range:
- 5 to 50°C (41 to 122°F)
- 5 to 50°C (41 to 122°F)