Erem® Magic™ 'N' Series All-Purpose Diagonal Cutters, Cooper Tools
Supplier: Cooper Tools
Certificates
About this item
These side cutters are including the features such as superior-grade tool steel construction, maximum opening stop, interlock screw joint system and ESD-safe cushion grips.
- Oval head is the most widely used head shape
- It is robust and size for size offers the highest cutting capacity
- Internal springs for fatigue-and wobble-free operation
- These cutters ensure high precision and extended life
The Cutting capacity varies based on the wire quality, these cutters are mainly suitable for cutting medium-hard wire, stainless steel wire, soft wire, copper and aluminum wires.