ContactUsLinkComponent
Magnetic Probe Holder for Positioning the PT100 Probes Easily in Refrigerators, Freezers, and Other Assets
Magnetic Probe Holder for Positioning the PT100 Probes Easily in Refrigerators, Freezers, and Other Assets
Catalog # 77394-758
Supplier:  SenseAnywhere
undefined
Magnetic Probe Holder for Positioning the PT100 Probes Easily in Refrigerators, Freezers, and Other Assets
Catalog # 77394-758
Supplier:  SenseAnywhere
Supplier Number:  0096647495422
Specifications
  • Description:
    The magnetic probe holder is perfect for positioning the PT100 probes easily in refrigerators, freezers, and other assets, boasting user-friendly installation
  • Cat. No.:
    77394-758
Specifications
About this item
The magnetic probe holder is perfect for positioning the Pt100 probes easily in refrigerators, freezers, and other assets, boasting user-friendly installation.
The hole diameter is precisely engineered between 3.1 to 3.2 mm to ensure a universal fit for both stainless steel and hermetically sealed probes, guaranteeing compatibility across a range of models.