About this item
The Nio™ digital PCR instrument line offers a simple all-in-one digital PCR workflow that supports seven colors, flexible throughput of up to 768 samples per day with three configurations, continuous loading, and easy-to-use software.
- All-in-one digital PCR platform
- Up to 7-color multiplexing
- Flexible throughput with three configurations: Nio™+, Nio™, and Nio™ E
- User-friendly design
- Continuous loading
- Remote access
- AI powered droplets count
Nio™ digital PCR system is a versatile, all-in-one, fit-for-throughput digital PCR instrument with uparalleled user-friendliness. Easy-to-use and highly automated, the Nio™ is the only digital PCR instrument that works exactly like laboratories work, featuring continuous sample loading capability, allowing the user to add up to eight chip plates whenever needed, and whether the instrument is running an experiment or not.
Available in three configurations to accommodate any lab's throughput needs. Nio™ harnesses Crystal Digital PCR® technology, which partitions samples into a large array of thousands of individual droplets - each its own reaction compartment - before amplifying nucleic acid molecules in each droplet. These reactions are tagged with fluorphores to be read using up to seven different fluorescence light channels.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77392-280
- 77449-124
- 77574-018
- Compatible With:
- FAM, Yakima Yellow®, Atto®550, ROX, Cy®5, Atto®700/Cy®5.5, DY-521-XL
- FAM, Yakima Yellow®, Atto®550, ROX, Cy®5, Atto®700/Cy®5.5, DY-521-XL
- FAM, Yakima Yellow®, Atto®550, ROX, Cy®5, Atto®700/Cy®5.5, DY-521-XL
- No. of Channels:
- 7 (Fluorescent channels)
- 7 (Fluorescent channels)
- 7 (Fluorescent channels)
- W×D×H:
- 56×65×52 cm
- 56×65×52 cm
- 56×65×52 cm
- Weight:
- 61 kg
- 61 kg
- 61 kg