Stilla® Sapphire Chip
Catalog # 77392-264
Supplier: STILLA TECHNOLOGIES, INC.
CAS Number:
Specifications
- Description:Sapphire chip
- Cat. No.:77392-264
- Units:Pack of 12 chips
Specifications
About this item
The Sapphire Chip features cutting-edge, patented microfluidic technology that integrates the entire digital PCR workflow into a single consumable.
- Capacity: Up to four samples/chip
- Input volume: 25 µl/chamber
- Droplets per sample: Up to 30000
- Dynamic range: ∼5 log
For maximum template input volume, the Sapphire Chip can be loaded with up to 4 samples per chip, and generate up to 30,000 droplets per sample for your digital PCR workflows. After sample preparation, just a single pipetting step is required before the chip is loaded onto either the 3-color naica® system or the 6-color naica® system for partitioning, PCR amplification, and sample read-out. The Sapphire Chip supports applications such as rare mutation detection, liquid biopsy analysis, gene expression analysis, and pathogen detection.