Kimble® KIMAX® Cream Pipettes with Wide Tip, DWK Life Sciences
Catalog # 48756-028
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Capacity:9 ml
- Cat. No.:48756-028
- Supplier no.:570-9
Specifications
About this item
Reusable glass pipettes with wide tip.
- Scale is permanent brown stain
- Calibrated to deliver
Designed from ASTM Specification E1043, Type III