Recombinant Coronavirus (SARS-CoV Envelope) Antigen (aa 1-76) (from E. coli)
76418-072
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Size:100 µg
- Storage Conditions:–20 °C
- Protein Synonyms:Coronavirus envelope protein
- Protein/Peptide Name:Coronavirus (SARS-CoV Envelope) antigen (aa 1-76)
- Molecular Weight:34 kDa
- Formulation:Liquid. In 50 mM Tris-HCl, 60 mM NaCl with 50% glycerol.
- Shipping Temperature:20
- Cat. No.:76418-072
Produced in E. coli. Coronavirus (SARS-CoV) envelope protein (aa 1-76).