Zika Virus Envelope Protein (Suriname Strain) (from HEK293 cells)
Catalog # 76418-052
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Conjugation:Unconjugated
- Source:HEK293 cells
- Size:100 µg
- Storage Conditions:–80 °C
- Protein/Peptide Name:Zika Virus Envelope protein (Suriname strain)
- Formulation:Liquid. In 20 mM NaH₂PO₄ pH 7.8, 210 mM NaCl
- Shipping Temperature:Dry Ice
- Tested Applications:Vaccine research, LINE assay, Antibody capture
- Cat. No.:76418-052
Specifications
About this item
Zika virus envelope protein recombinant antigen containing a C-terminal His-tag.