Human Recombinant Nampt (from E. coli)
76418-048
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 µg
- Storage Conditions:–80 °C
- Endotoxin Content:<1 EU/µg protein (LAL test)
- Reconstitution Instructions:Reconstitute in sterile 20 mM HCl at 0.1 mg/ml
- Protein Synonyms:Nicotinamide phosphoribosyltransferase|Pre-B-cell colony enhancing factor 1|Visfatin|PBEF
- Protein/Peptide Name:Nampt
- Molecular Weight:∼52.7 kDa
- Formulation:Lyophilized from a sterile (0.2 µm) filtered aqueous solution containing 0.1% Trifluoroacetic Acid (TFA)
- Shipping Temperature:Ambient temperature
- Cat. No.:76418-048
Produced in E. coli. Contains 466 amino acids.