ContactUsLinkComponent
Spor-Klenz® Ready-to-Use Disinfectant Wipes
  76325-902
 :  Steris
undefined
Spor-Klenz® Ready-to-Use Disinfectant Wipes
  76325-902
 :  Steris
 :  652915
  • Sterility:
    Sterile
  • Color:
    White
  • Cat. No.:
    76325-902
  • Packaging:
    Double bagged
  • W×L:
    22.9×22.9 cm (9×9")
Spor-Klenz® ready-to-use disinfectant wipes are specifically formulated for convenient disinfection of hard surfaces.
  • Eliminates the need to validate dilution preparation procedures by providing saturated wipes with minimal operator effort
  • Saves time and labor of chemical preparation
  • Microbial control against a broad spectrum of bacteria, fungi, and viruses
  • Low-linting wipes with sealed edges for reduced particle generation
  • Gamma irradiated for easy introduction into aseptic areas
  • Same formula as Spor-Klenz® ready-to-use
  • Each lot is USP sterility tested and documented by a certificate of analysis
  • Manufactured in accordance with the cGMP with complete lot traceability
Spor-Klenz® ready-to-use disinfectant wipes are formulated for use on hard, non-porous surfaces. Made with the Spor-Klenz® ready-to-use formula, the Spor-Klenz® RTU dsinfectant wipes kill a broad spectrum of common bacteria, fungi, and viruses. The wipes use an innovative system that combines woven polyester wipes with sealed edges, packaged with a separate internal pouch of Spor-Klenz® RTU formula. This internal pouch allows for maximum chemical stability while offering a simple, convenient, and consistent method to saturate the wipes. By simply rolling the package of wipes, the internal pouch of liquid opens and quickly saturates the wipes.
Each package is double-bagged and gamma irradiated for use in aseptic areas.
 : Certificates of manufacture/analysis for consumable products by product ID and lot number.
 : Wipes are packaged 12 pouches per case (180 wipes).