Alnochromix® Oxidizing Acid Additives for Glass Cleaning
: Alconox
Concentrated inorganic persulfate based additive to sulfuric acid to make a metal-free and surfactant-free glassware cleaner. It is a non metallic, non-carcinogenic, and easier to dispose replacement for chromic acid.
- Metal-free for trace metal glassware cleaning
- Surfactant-free for enzyme kinetics, cell and tissue culture cleaning
- Removes stubborn odors from containers, difficult organic, cyanide and metal residues
- Replaces carcinogenic and hazardous chromic acid
- Free rinsing to provide reliable results and no interfering residues
- Two years shelf life
These can be used to clean laboratory glassware and other glass, and ceramic surfaces used for trace metal, enzyme kinetics, flavor profile, tissue and cell culture procedures.
Recommended for cleaning glass, ceramic and acid resistant plastic surfaces, using soak or brush method.
For best results use freshly made solution within first seven days of mixing. When cleaning solution is stale or discolored, it can be regenerated by adding new Alnochromix®.
Color: White
: Do not close solution container airtight, use vented caps (76322-110). Never use nitric acid in place of sulfuric acid. Do not mix silver salt, glassware containing silver salts should be rinsed before soaking in Alnochromix® and sulfuric acid solution. Wear suitable personal protective equipment for skin and eyes. Silver gloves (76322-112) available.