ContactUsLinkComponent
Automation Tips for Tecan Fluent and Evo Workstations
  MSPP-LHA-50R-F
undefined
Automation Tips for Tecan Fluent and Evo Workstations
  MSPP-LHA-50R-F
 :  LHA-50R.F
  • Volume:
    50 µl
  • Tip color:
    Clear
  • Packed:
    96 Tips/Rack, 8 Racks/Pack, 3 Packs/case
  • Cat. No.:
    MSPP-LHA-50R-F
  • Filter:
    +
  • Sterile:
    +
  • Tip style:
    Tecan fluent Evo LiHa compatible P50 - single stack, refill tray
  • For use with:
    Tecan fluent and Evo LiHa/FCA heads
Fluotics manufactures a complete selection of automation tips compatible with Tecan fluent and Evo workstations. These automation tips are directly interchangeable with their Tecan automation tip counterpart, with no change to the labware definitions required.
  • Certified RNase-/DNase-free and nonpyrogenic
  • Produced from virgin, high-grade polypropylene
  • Available in non sterile, sterile, and barrier filter formats
Compatible with Tecan fluent and Evo workstations with MCA and LiHA/FCA heads. Ultra polishing techniques ensure low liquid retention without the use of releasing agents or other additives.