Automation Tips for Beckman Coulter® Biomek® i-Series, FX/NX and Multimek Workstations
Supplier: FLUOTICS INC MS
Certificates
About this item
Fluotics manufactures a selection of automation tips compatible with Beckman Coulter® Biomek® i-Series, FX/NX, and Multimek workstations. These automation tips are directly interchangeable with their Beckman automation tip counterpart, with no change to the labware definitions required.
- Certified RNase-/DNase-free and nonpyrogenic
- Produced from virgin, high grade polypropylene
- Available in non sterile, sterile and barrier filter formats
Compatible with Biomek® i-Series, FX/NX and Multimek workstations with 384 heads. Ultra polishing techniques ensure low liquid retention without the use of releasing agents or other additives.
Packaging: Pipette tips are packaged 384 per rack, 50 racks per case.