Linearity FD Special Diabetes
Catalog # 77391-768
Supplier: Audit Microcontrols
CAS Number:
Specifications
- Description:Linearity FD special diabetes
- Cat. No.:77391-768
- Volume:2 ml
Specifications
About this item
Linearity FD special diabetes is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analyte: C-peptide and Insulin.
- Linearity
- Freeze dried