Three-sided Ceiling Kit for Protector® XL and XStream Laboratory Hoods
89370-046
: Labconco
Product available on GSA Advantage®
- Description:Ceiling kit
- Height:18.6 - 24.4"
- Cat. No.:89370-046
- Depth:37.7"
- Width:8 ft.
- Supplier no.:9414812
Ceiling enclosure kits include panels that extend above the top of the hood to hide exposed ductwork, plumbing and wiring.
- Can be used with Protector Premier, XStream and XL laboratory hoods
- Glacier white powder-coated steel
These three-sided ceiling enclosure kits include three glacier white powder-coated steel ceiling enclosure panels, one for the front of the hood and one for each side of the hood.