Self-Sealing Sterilization Peel Pouches, Medical Paper/Film, with Steam Indicator
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
New Product
Certificates
About this item
Designed for steam sterilization cycles (121 to 135 °C).
- Peels open cleanly
- Triple weld factory seals
- Convenient, time-saving self-seal closure system
- Strong, rip- and tear-resistant 60 g/m² medical grade paper material
- Good barrier characteristics prevent microbial penetration
- Sub-packed into sealed labeled poly bags with manufacturing lot number
- Date of manufacture on label for complete traceability
Significant improvement versus cutting and wrapping with sheets and autoclave tape.
Certifications: Certificates of Conformance sent with each shipment.