ContactUsLinkComponent
AssetTag for uninterrupted Surveillance and Data during Calibration
AssetTag for uninterrupted Surveillance and Data during Calibration
Catalog # 77391-864
Supplier:  SenseAnywhere
undefined
AssetTag for uninterrupted Surveillance and Data during Calibration
Catalog # 77391-864
Supplier:  SenseAnywhere
Supplier Number:  0096647494982
Specifications
  • Description:
    AssetTags for uninterrupted surveillance and data during calibration
  • Cat. No.:
    77391-864
Specifications
About this item
To maintain quality standards, industries must calibrate sensors regularly, potentially disrupting monitoring and creating data gaps. SenseAnywhere's AssetTagging system addresses this by enabling continuous surveillance with interchangeable data loggers. This approach ensures no data gaps during calibration and eliminates the need to reset alarms and settings.
  • Monitor temp/RH of the asset
  • Data loggers can be switched at any time
  • Complete temp/RH history of assets without any monitoring-gaps
  • No reconfiguration needed (alarms, reports etc.)
  • No paperwork or administration
  • No prone to error or hassle
  • Complete audit trail
The AssetTag feature simplifies the process of swapping AiroSensors, ensuring data continuity. The AssetTag is equipped with a QR-code, which upon scanning, facilitates the transition to another AiroSensor for ongoing measurements. This ease of exchange is particularly useful for AiroSensors requiring recalibration. Importantly, all previously established alarms and settings automatically apply to the new AiroSensor, eliminating the need for reconfiguration. As a result, monitoring remains uninterrupted, and data loggers can be switched seamlessly at any time, maintaining the integrity and consistency of your surveillance system.
Ordering information: Explore the full range of SenseAnywhere products and services by visiting the Documentation tab, selecting 'Additional Product Details,' and opening the 'Which SenseAnywhere Data Logger Do I Need' document.