ContactUsLinkComponent
Staticide® 7060 Polyester Swab
Staticide® 7060 Polyester Swab
Catalog # 77200-552
Supplier:  ACL Staticide
undefined
Staticide® 7060 Polyester Swab
Catalog # 77200-552
Supplier:  ACL Staticide
Supplier Number:  7060
Specifications
  • Length:
    6.3"
  • Material:
    Medical-grade foam
  • Cat. No.:
    77200-552
  • Tip style:
    Narrow paddlehead
Specifications
About this item
Robust and reliable knit swab for cleanroom applications.
  • Non-abrasive, double-layer polyester knit tip
  • Ultra-low particle; lint-free
  • Free of silicon, amides, and DOP contamination
  • White polypropylene handles
  • Class 10 laundered and packaged
  • ISO Class 5 suitable
The swab construction features ultra sonic welding, with no adhesives or binders utilized.