Staticide® 8150 Economy ESD Wrist Strap Kit, 4 mm
Catalog # 77200-604
Supplier: ACL Staticide
Specifications
- Color:Dark blue
- Description:Economy ESD wrist strap kit, 4 mm
- Environmentally Preferable:
- Cat. No.:77200-604
Specifications
About this item
Adjustable economy wrist strap kit offers comfortable and secure personal grounding for occasional use.
- Includes dark blue wrist strap, cord, and alligator clip
- Recommended for occasional wear
- Elastic crochet fabric linded with 5 rows of double-stitched silver yarn
- 1.5 mm band thickness
Once connected to ground, the wrist strap provides a safe path for eliminating static charges while working on electronics.