ContactUsLinkComponent
Staticide® Ground Cord for ESD Mats, 10 mm Stud Snap
Staticide® Ground Cord for ESD Mats, 10 mm Stud Snap
Catalog # 77200-568
Supplier:  ACL Staticide
undefined
Staticide® Ground Cord for ESD Mats, 10 mm Stud Snap
Catalog # 77200-568
Supplier:  ACL Staticide
Supplier Number:  8090
Specifications
  • Description:
    Ground cord for ESD mats
  • Environmentally Preferable:
  • Cat. No.:
    77200-568
Specifications
About this item
The 8090 Mat ground cord provides a simple connection between the mat and ground source.
  • 10' coated high-strength tensile wire
  • 1-meg ohm resistor
  • 10 mm male stud connection to mats
  • Ring-style termination for connecting to grounding post or bar