ContactUsLinkComponent
Staticide® IPA Aerosol
Staticide® IPA Aerosol
Catalog # 77200-606
Supplier:  ACL Staticide
undefined
Staticide® IPA Aerosol
Catalog # 77200-606
Supplier:  ACL Staticide
Supplier Number:  8625
Specifications
  • Description:
    IPA aerosol
  • Cat. No.:
    77200-606
Specifications
About this item
Ideal choice for general cleaning applications on a variety of substrates where removing contaminants are of concern.
  • Flammable
  • 99.8% pure isopropyl alcohol
  • Made in the USA
Extra-strength IPA aerosol is safe for use on plastics. Evaporates quickly, dries fast, and leaves no residue.