Staticide® 7022 Polyurethane Foam Swab
Catalog # 77200-550
Supplier: ACL Staticide
Specifications
- Length:5.04"
- Material:Medical-grade foam
- Cat. No.:77200-550
- Tip style:Large paddlehead
Specifications
About this item
Medical-grade foam tipped swab with superior solvent capacity.
- High solvent capacity
- Non-abrasive medical-grade foam
- White polypropylene handles
- Class 10 laundered and packaged
- ISO Class 4-7 compatible
The swab construction features ultra sonic welding, with no adhesives or binders utilized.