ContactUsLinkComponent
Staticide® 8122 Premium Heel Grounder
Staticide® 8122 Premium Heel Grounder
  77200-600
undefined
Staticide® 8122 Premium Heel Grounder
  77200-600
 :  8122
  • Color:
    Black
  • Description:
    Premium heel grounder
  • Cat. No.:
    77200-600
The premium heel grounder provides added conductivity, strength and longevity thanks to an extra-wide grounding strap.
  • Two-layer heel rubber
  • Wide cup-style
  • Reinforced with steel rivets
  • 1-meg ohm resistor
  • Black conductive exterior: 10⁴ ohms
This premium heel grounder features a conductive ground ribbon measuring 14.5×0.05", and can be secured with an adjustable hook and loop fabric. It is specifically designed for use by grounding employees on the move in electronic manufacturing areas.