Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Closures
Test/Sample Tube Closures
Screw Caps for 30 ml Bulk Tubes, Caplugs
Screw Caps for 30 ml Bulk Tubes, Caplugs
Screw Caps for 30 ml Bulk Tubes, Caplugs
Catalog # MSPP-33532G2E1
Supplier:
CAPLUGS MS
Expand image
Screw Caps for 30 ml Bulk Tubes, Caplugs
Catalog # MSPP-33532G2E1
Supplier:
CAPLUGS MS
Supplier Number:
300-3532-G20
Specifications
Color:
Green
Description:
Screw cap
Cat. No.:
MSPP-33532G2E1
Packaging:
Bulk
For tubes:
20/30 ml
Specifications
About this item
Product Details
Product Details