Order Entry
Puerto Rico
ContactUsLinkComponent
Diluent B (with rAlbumin)
Diluent B (with rAlbumin)
Catalog # 77322-304
CAS Number:  
undefined
Diluent B (with rAlbumin)
Catalog # 77322-304
Supplier Number:  B8533S
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Concentration:
    1X
  • Description:
    Diluent B (with rAlbumin)
  • Size:
    5 ml
  • Cat. No.:
    77322-304

Specifications

About this item

Diluent B (with rAlbumin) is recommended for making dilutions of New England Biolabs, Inc. restriction enzymes.

  • Contains 4×1.25 ml vials
  • Restriction enzymes should be stored at their selling concentrations
  • Contains rAlbumin

Diluent B (with rAlbumin) is recommended for making dilutions of some NEB restriction enzymes. When necessary, we recommend diluting enzymes just prior to use and suggest that the final concentration of diluted enzymes be at least 1000 units/ml. Restriction enzymes should be stored at their selling concentrations.