NEBNext® UltraExpress™ DNA Library Prep Kits
The NEBNext® UltraExpress™ DNA library preparation kit is the latest generation of NEBNext® DNA library prep, with a fast, streamlined workflow.
- Fast workflow (<2 h)
- Fewer steps and consumables
- Fewer cleanups
- Single protocol for all input amounts
- High-quality libraries from a wide input range (10 to 200 ng pre-sheared DNA)
In under 2 h, the protocol enables the creation of high yield, high-quality libraries, from a broad input amount range, while generating less plastic waste. Further, with simplicity at the forefront, the kit features a single protocol for all input amounts, making NEBNext® UltraExpress™ DNA library prep well suited for all throughputs.
Frequently Bought Together