- Size:200 units
- Cat. No.:77322-322
A thermostable homolog of the E. coli Uracil-DNA Glycosylase (UDG) (1,2) from Archaeoglobus fulgidus.
- Archaeoglobus fulgidus Uracil DNA Glyocsylase
- Thermostable DNA glycosylase that hydrolyzes deoxyuridine to release uracil
- WarmStart® technology inhibits enzyme activity below 37 °C facilitating post-SDA carry-over prevention
- Active on ss and dsDNA
- Heat inactivation not possible
WarmStart® Afu UDG catalyzes the release of free uracil from uracil-containing DNA. It is formulated with a reversibly-bound aptamer, which inhibits its activity at temperatures below 37 °C. WarmStart® Afu UDG efficiently hydrolyzes uracil from single-stranded or double-stranded DNA.