Order Entry
Puerto Rico
ContactUsLinkComponent
 
Vacuum Cleaner, Dry Recovery-Only, CR-1000 (CV)
  77008-230
undefined
Vacuum Cleaner, Dry Recovery-Only, CR-1000 (CV)
  77008-230
 :  111994A

 

  • Airflow:
    96 CFM
  • Vacuum Pressure:
    99" H2O
  • Capacity:
    2 gal.
  • Description:
    Vacuum, pharma, dry recovery only with safe containment canister for potent carcinogenic powders
  • Model:
    CR-1000 (CV)
  • W×D×H:
    16×23×26"
  • Horsepower:
    1.3 HP
  • Current:
    8.3 A
  • Frequency:
    60 Hz
  • Noise level:
    60 dB(A)
  • Power:
    1000 W
  • Voltage:
    120 V
  • Weight:
    24 lbs.
  • Cat. No.:
    77008-230

 

 

ULPA filtered cleanroom vacuum designed for the recovery of potent powders in hazardous locations. Safe containment canister for the safe collection, containment and disposal of potent compounds included.

  • Cleanroom compatible with ISO Class 4 (former Class 10); can be used in the pharmaceutical industry
  • Five-stage filtration system, includes an ULPA filter (U15), HEPA filter (H14), PTFE laminated main cloth filter, safety filter, and disposable filter bags
  • Designed for the recovery of potent powders in hazardous locations
  • Safe containment canister for the safe collection, containment and disposal of potent compounds included
  • Thru-Flow heavy duty motor
  • Low noise level - only 60 dB(A) at 6.6' (2 m)
  • For dry recovery only
  • Autoclavable stainless steel Type 316 filter chamber and wheel base
  • Mounted on a four-wheel dolly for easy maneuverability
  • Three optional tool kits (not included) - standard, ESD safe, and autoclavable

ULPA filter with an efficiency of 99.999% at 0.12 micron. Filtration efficiency of 99.9995% at 0.18 micron. Tested IEST-RP-CC001. U15 by MPPS method as per EN 1822. All of our ULPA vacuum systems are aerosol leak tested before leaving our facility - included.

HEPA filter with an efficiency of 99.995% at 0.3 micron. Tested IEST-RP-CC001. H14 by MPPS method as per EN 1822 and OSHA compliant. All of our HEPA vacuum systems are aerosol leak tested before leaving our facility - included.

Suction inlet: 1.5"

 : Standard(s) for Electric Vacuum Cleaning Machines and Blower Cleaners (UL-1017).
 : For available options, accessories, or tool kits (not included); contact your Avantor representative.