Order Entry
Puerto Rico
ContactUsLinkComponent
Vacuum Cleaner, Dry Recovery-Only, C-10 EX (4W)
Vacuum Cleaner, Dry Recovery-Only, C-10 EX (4W)
Catalog # 77008-228
undefined
Vacuum Cleaner, Dry Recovery-Only, C-10 EX (4W)
Catalog # 77008-228
Supplier Number:  111805A

Specifications

  • Airflow:
    110 CFM
  • Vacuum Pressure:
    107" H2O
  • Capacity:
    5 gal.
  • Description:
    Vacuum, dry recovery only
  • Model:
    C-10 EX (4W)
  • W×D×H:
    17×17×36.5"
  • Horsepower:
    1.6 HP
  • Current:
    10.5 A
  • Frequency:
    60 Hz
  • Noise level:
    72 dB(A)
  • Power:
    1207 W
  • Voltage:
    120 V
  • Weight:
    52 lbs.
  • Cat. No.:
    77008-228

Specifications

About this item

HEPA filtered vacuum used to recover combustible/conductive dusts in Hazardous Locations Division 2. Excellent seller in addititve manufacturing and 3D printing.

  • Ideal for housekeeping and general cleaning
  • Includes four HEPA filters, Class H14, individually factory tested
  • Heavy duty two-stage by-pass motor
  • Longer life brushless motor available; always with 4×H14 HEPA filters
  • Material to be recovered within a conductive cloth recovery bag
  • Includes an indicator light to warn of vacuum blockage
  • Includes a complete static dissipative tool kit

Recovery tank made of stainless steel Type 430 (recovery tank made of stainless steel Type 304 is also available).

Suction inlet: 1.5"

Certifications: Dust ignition protected. Cleaning machines for Hazardous Locations (Classes C1618 01 & C1618 81). Division 2, Class II, Groups F & G.
Ordering information: For available options or accessories; contact your Avantor representative.