Virtu-Clean® Disposable Cleaning Pads
Catalog # 77007-932
Supplier: High-Tech Conversions
Specifications
- Description:Virtu-Clean® disposable cleaning pad
- Packed:20 pads/bag, 12 bags/master bag
- Cat. No.:77007-932
Specifications
About this item
Not all cleaning pads are equal. Absorbency rates, release rates, coverage, and linting can vary greatly, significantly impacting the finished product.
- Cleanroom class: ISO 6 (Class 1000)
- 50% post-consumer recycled content
- Eco friendly manufacturing with no chemical dyes used in the process
- Single-use application to reduce risk of cross contamination
- Embossed pattern reduces friction for less strain on workers
- Channels efficiently pick up and retain dirt in pad
- No Quat binding
- Compatible with chlorine bleach, hydrogen peroxide, and common disinfectants
- 300 to 360 sq. ft. cleaning coverage
- Absorbs 9X its weight in liquid
Superior absorbency and good release provide consistent coverage and cleaning, Bonded edges improve performance greatly, reducing linting for cleaner results. Bag in bag packaging for cleanroom application. Perfect for foodservice and food processing operations, healthcare facilities, cleanrooms, controlled environments, surgical rooms, ICU areas, pharmacy applications, and other institutions.