ContactUsLinkComponent
Drosophila Olfactory Operant Conditioning Apparatus
Drosophila Olfactory Operant Conditioning Apparatus
Catalog # 76670-412
Supplier:  MAZE ENGINEERS
undefined
Drosophila Olfactory Operant Conditioning Apparatus
Catalog # 76670-412
Supplier:  MAZE ENGINEERS
Supplier Number:  8501
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Drosophila olfactory operant conditioning apparatus
  • Environmentally Preferable:
  • Cat. No.:
    76670-412
Specifications
About this item
The Drosophila Olfactory Operant Conditioning is an odor-aversion learning apparatus for Drosophila.
  • 2 odor chambers
  • T-Maze choice area
  • Copper testing tube
  • Vacuum lines
  • Secure clamp
The Drosophila Olfactory Operant Conditioning apparatus was first developed by Tully and Quinn (Tully and Quinn, 1985) using a combination of a T-Maze electric-shock reinforcement in a copper wire grid. The unit includes a clear, robust apparatus that includes a holding area and T-Maze choice areas leading into odor compartments.