ContactUsLinkComponent
Drosophila Maze Array
Drosophila Maze Array
Catalog # 76670-416
Supplier:  MAZE ENGINEERS
undefined
Drosophila Maze Array
Catalog # 76670-416
Supplier:  MAZE ENGINEERS
Supplier Number:  8505
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Drosophila maze array (25 units)
  • Material:
    Acrylic
  • Environmentally Preferable:
  • Cat. No.:
    76670-416
Specifications
About this item
The Drosophila Maze Array is designed to study locomotor behavior in Drosophila.
  • Each array is well lit in this configuration
  • Multiple arrays possible including Y, T, +, and -
  • Customized array shapes are available upon request
Locomotor behavior is a form of behavioral handedness, or the preferential performance of behavior on one side of the body or with a particular chiral twist (Buchanan, 2015). This locomotor handedness is not related to a genetic dominance or morphological asymmetry. Instead, it is related to the asymmetric involved in the collection and processing of sensory information by the subject. In the Drosophila Maze Array, light and different pattern configurations are used to study how specific sensory exposures affect locomotive behavior.
The Y-Maze Array configuration is an efficient test to evaluate the chemosensory responses in Drosophila (Simonnet, 2014). The T-Maze Array configuration can be utilized to test memory (Malik, 2014) and phototaxic reflex in adult flies (Swinderen, 2011). By studying Drosophila locomotor behavior using the different maze array configurations, one can start to understand and analyze the variability in physiological, morphological, and behaviors exhibited by Drosophila that have grown up in identical environments (Buchanan, 2015).