Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Width:8 cm
- Description:Automated zebrafish Y-maze
- Height:15 cm
- Length:25 cm
- Material:Acrylic
- Cat. No.:76670-134
Specifications
About this item
The automated avoidance Zebrafish Y-Maze is an operant conditioning assay used to study avoidance behaviors in Zebrafish when presented with an aversive stimulus.
- LCD screen placed below maze displays visual cues throughout all maze regions
- Shock and sound stimuli also available
- Custom coloring and customization available
The Automated Avoidance Zebrafish Y-Maze consists of a Y-Maze with an LCD screen placed below the maze that displays visual cues throughout all regions of the maze.